Assay Details
Target Gene Details
Entrez Gene ID: | 101060264 |
Gene Name: | FOXP4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:41523895-41548621 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXP4-AS1 | NR_126415.1 | |||
| NR_126416.1 | ||||
| NR_126417.1 | ||||
| BF194903.1 | ||||
| BG028463.1 | ||||
| BQ082169.1 | ||||
| BQ128057.1 | ||||
| BQ581966.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79037960] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map