Assay Details
Target Gene Details
Entrez Gene ID: | 135293 |
Gene Name: | peptidase M20 domain containing 2 |
Gene Aliases: |
ACY1L2, bA63L7.3 |
Location: |
Chr.6:89096879-89165569 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PM20D2 | XM_005248662.4 | XP_005248719.1 | ||
| XM_011535481.2 | XP_011533783.1 | |||
| XM_017010291.1 | XP_016865780.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023532 | Chr.6:89093252 - 89253488 on Build GRCh38 | Loss |
|
| nsv1027007 | Chr.6:89099218 - 89628146 on Build GRCh38 | Gain |
|
| esv3609896 | Chr.6:89132295 - 89137224 on Build GRCh38 | Loss |
|
| esv2662579 | Chr.6:89132296 - 89137220 on Build GRCh38 | Deletion |
|
| esv23341 | Chr.6:89132231 - 89137224 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map