Assay Details
Target Gene Details
Entrez Gene ID: | 26002 |
Gene Name: | monooxygenase DBH like 1 |
Gene Aliases: |
MOX, PRO5780, dJ248E1.1 |
Location: |
Chr.6:132296055-132401534 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOXD1 | NM_015529.3 | 12 | 2656 | NP_056344.2 |
| XM_017010714.1 | 12 | 2464 | XP_016866203.1 | |
| XM_017010715.1 | 12 | 2302 | XP_016866204.1 | |
| AJ420565.1 | 2 | 916 | ||
| AK074879.1 | 12 | 2371 | BAC11263.1 | |
| AK226054.1 | 8 | 1753 | ||
| AL080058.1 | 8 | 1714 | CAB45692.1 | |
| AY007239.1 | 11 | 2572 | AAG09636.1 |
More Information
Set Membership: |
|

Genomic Map