Assay Details
Target Gene Details
Entrez Gene ID: | 285780 |
Gene Name: | LY86 antisense RNA 1 |
Gene Aliases: |
LY86-AS, LY86AS |
Location: |
Chr.6:6346465-6622826 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LY86-AS1 | NR_026970.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470796 | Chr.6:6341947 - 6368956 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76433530] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map