Assay Details
Target Gene Details
Entrez Gene ID: | 5603 |
Gene Name: | mitogen-activated protein kinase 13 |
Gene Aliases: |
MAPK 13, MAPK-13, PRKM13, SAPK4, p38delta |
Location: |
Chr.6:36130484-36144524 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK13 | NM_002754.4 | 12 | 3720 | NP_002745.1 |
| NR_072996.1 | 10 | 3568 |
More Information
Additional Information:
For this assay, SNP(s) [rs73730464] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map