Assay Details
Target Gene Details
Entrez Gene ID: | 3710 |
Gene Name: | inositol 1,4,5-trisphosphate receptor type 3 |
Gene Aliases: |
IP3R, IP3R3 |
Location: |
Chr.6:33621333-33696574 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITPR3 | NM_002224.3 | NP_002215.2 | ||
| XM_017010832.1 | XP_016866321.1 | |||
| BC146646.1 | ||||
| D26351.1 | BAA05385.1 | |||
| U01062.1 | AAC50064.1 |
Target Gene Details
Entrez Gene ID: | 101929188 |
Gene Name: | uncharacterized LOC101929188 |
Gene Aliases: |
- |
Location: |
Chr.6:33631244-33633745 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929188 | NR_134628.1 | 2 | 730 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608630 | Chr.6:33439052 - 34076935 on Build GRCh38 | Gain |
|
| dgv85n68 | Chr.6:33583425 - 33743569 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map