Assay Details
Target Gene Details
Entrez Gene ID: | 9450 |
Gene Name: | lymphocyte antigen 86 |
Gene Aliases: |
MD-1, MD1, MMD-1, dJ80N2.1 |
Location: |
Chr.6:6588639-6654983 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LY86 | NM_004271.3 | NP_004262.1 | ||
| XM_017011505.1 | XP_016866994.1 | |||
| AF057178.2 | AAC98152.2 | |||
| AY931019.1 | ||||
| BC038846.1 | AAH38846.1 | |||
| CD695164.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607998 | Chr.6:6603300 - 6729633 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map