Assay Details
Target Gene Details
Entrez Gene ID: | 2940 |
Gene Name: | glutathione S-transferase alpha 3 |
Gene Aliases: |
GSTA3-3, GTA3 |
Location: |
Chr.6:52896639-52909799 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSTA3 | NM_000847.4 | NP_000838.3 | ||
| XM_006715072.3 | XP_006715135.1 | |||
| AF020919.1 | AAD04712.1 | |||
| BC020619.1 | AAH20619.1 | |||
| DT932587.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3609013 | Chr.6:52901914 - 52922345 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map