Assay Details
Target Gene Details
Entrez Gene ID: | 91749 |
Gene Name: | major facilitator superfamily domain containing 4B |
Gene Aliases: |
KIAA1919, NaGLT1 |
Location: |
Chr.6:111259279-111280005 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD4B |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1020627 | Chr.6:110982539 - 111302511 on Build GRCh38 | Gain |
|
| nsv1028349 | Chr.6:111184772 - 111371598 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140615185] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map