Assay Details
Target Gene Details
Entrez Gene ID: | 81696 |
Gene Name: | olfactory receptor family 5 subfamily V member 1 |
Gene Aliases: |
6M1-21, hs6M1-21 |
Location: |
Chr.6:29355211-29457067 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR5V1 | XM_011514929.2 | XP_011513231.1 | ||
| XM_011514930.2 | XP_011513232.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1145096 | Chr.6:28626523 - 29718723 on Build GRCh38 | Deletion |
|
| nsv1074389 | Chr.6:29116522 - 31446623 on Build GRCh38 | Deletion |
|
| nsv830624 | Chr.6:29306748 - 29477788 on Build GRCh38 | Loss |
|
| nsv1122126 | Chr.6:28628023 - 31244023 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115056830] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map