Assay Details
Target Gene Details
Entrez Gene ID: | 2914 |
Gene Name: | glutamate metabotropic receptor 4 |
Gene Aliases: |
GPRC1D, MGLUR4, mGlu4 |
Location: |
Chr.6:34018643-34155622 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRM4 | NM_000841.3 | 11 | 6136 | NP_000832.1 |
| NM_001256809.2 | 9 | 5519 | NP_001243738.1 | |
| NM_001256811.2 | 10 | 5995 | NP_001243740.1 | |
| NM_001256812.2 | 10 | 5220 | NP_001243741.1 | |
| NM_001256813.2 | 11 | 5444 | NP_001243742.1 | |
| NM_001282847.1 | 10 | 5919 | NP_001269776.1 | |
| XM_017010790.1 | 11 | 6074 | XP_016866279.1 | |
| XM_017010791.1 | 11 | 6129 | XP_016866280.1 | |
| XM_017010792.1 | 10 | 5673 | XP_016866281.1 | |
| XM_017010793.1 | 10 | 5424 | XP_016866282.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608630 | Chr.6:33439052 - 34076935 on Build GRCh38 | Gain |
|
| nsv602861 | Chr.6:33985869 - 34083140 on Build GRCh38 | Loss |
|
| nsv602843 | Chr.6:33964665 - 34227234 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map