Assay Details
Target Gene Details
Entrez Gene ID: | 8899 |
Gene Name: | pre-mRNA processing factor 4B |
Gene Aliases: |
PR4H, PRP4, PRP4H, PRP4K, dJ1013A10.1 |
Location: |
Chr.6:4019402-4064983 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1140598 | Chr.6:4020778 - 4020828 on Build GRCh38 | Deletion |
|
| nsv1074366 | Chr.6:4020777 - 4020828 on Build GRCh38 | Deletion |
|
| esv2731499 | Chr.6:4020698 - 4020988 on Build GRCh38 | Deletion |
|
| esv2370815 | Chr.6:4020581 - 4021022 on Build GRCh38 | Deletion |
|
| esv2493637 | Chr.6:4020781 - 4020830 on Build GRCh38 | Deletion |
|
| esv1658451 | Chr.6:4020780 - 4020830 on Build GRCh38 | Deletion |
|
| esv2731500 | Chr.6:4020812 - 4020935 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs567961417] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map