Assay Details
Target Gene Details
Entrez Gene ID: | 2940 |
Gene Name: | glutathione S-transferase alpha 3 |
Gene Aliases: |
GSTA3-3, GTA3 |
Location: |
Chr.6:52896639-52909799 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSTA3 | NM_000847.4 | NP_000838.3 | ||
| XM_006715072.3 | XP_006715135.1 | |||
| XM_011514533.2 | XP_011512835.1 | |||
| XM_017010794.1 | XP_016866283.1 | |||
| AA846513.1 | ||||
| AF020919.1 | AAD04712.1 | |||
| AF508266.1 | AAM33360.1 | |||
| AI199811.1 | ||||
| BC020619.1 | AAH20619.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv970123 | Chr.6:52894055 - 52905719 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map