Assay Details
Target Gene Details
Entrez Gene ID: | 221468 |
Gene Name: | transmembrane protein 217 |
Gene Aliases: |
C6orf128, dJ355M6.2 |
Location: |
Chr.6:37212178-37259034 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM217 | NM_001162900.1 | NP_001156372.1 | ||
| NM_001286401.1 | NP_001273330.1 | |||
| NM_145316.3 | NP_660359.2 | |||
| NR_104442.1 | ||||
| NR_104443.1 | ||||
| XM_011514362.1 | XP_011512664.1 | |||
| XM_011514363.2 | XP_011512665.1 | |||
| XM_011514364.2 | XP_011512666.1 | |||
| XM_011514365.2 | XP_011512667.1 | |||
| XM_011514366.2 | XP_011512668.1 | |||
| XM_011514367.2 | XP_011512669.1 | |||
| XM_011514369.2 | XP_011512671.1 | |||
| AK098666.1 | BAC05371.1 | |||
| BC026012.1 | AAH26012.1 | |||
| BC029657.1 | AAH29657.1 | |||
| BG285024.1 | ||||
| DB458508.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|

Genomic Map