Assay Details
Target Gene Details
Entrez Gene ID: | 60685 |
Gene Name: | zinc finger AN1-type containing 3 |
Gene Aliases: |
TEX27 |
Location: |
Chr.6:37819496-38154624 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFAND3 | NM_021943.2 | NP_068762.1 | ||
| XM_005249269.1 | XP_005249326.1 | |||
| XM_011514790.1 | XP_011513092.1 | |||
| AI817165.1 | ||||
| AK023284.1 | BAB14508.1 | |||
| BC031481.2 | AAH31481.1 | |||
| BP305887.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608721 | Chr.6:37819657 - 37855416 on Build GRCh38 | Gain |
|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76767526] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map