Assay Details
Target Gene Details
Entrez Gene ID: | 8899 |
Gene Name: | pre-mRNA processing factor 4B |
Gene Aliases: |
PR4H, PRP4, PRP4H, PRP4K, dJ1013A10.1 |
Location: |
Chr.6:4019402-4064983 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv349483 | Chr.6:4029034 - 4029346 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114754834] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map