Assay Details
Target Gene Details
Entrez Gene ID: | 7422 |
Gene Name: | vascular endothelial growth factor A |
Gene Aliases: |
MVCD1, VEGF, VPF |
Location: |
Chr.6:43770209-43786487 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
More Information
Set Membership: |
|

Genomic Map