Assay Details
Target Gene Details
Entrez Gene ID: | 83741 |
Gene Name: | transcription factor AP-2 delta |
Gene Aliases: |
TFAP2BL1 |
Location: |
Chr.6:50713544-50773033 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFAP2D | NM_172238.3 | NP_758438.2 | ||
| AY028376.1 | AAK16926.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74388152] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map