Assay Details
Target Gene Details
Entrez Gene ID: | 2309 |
Gene Name: | forkhead box O3 |
Gene Aliases: |
AF6q21, FKHRL1, FKHRL1P2, FOXO2, FOXO3A |
Location: |
Chr.6:108559823-108684769 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXO3 | NM_001455.3 | NP_001446.1 | ||
| NM_201559.2 | NP_963853.1 | |||
| XM_005266867.4 | XP_005266924.1 | |||
| XM_011535626.1 | XP_011533928.1 | |||
| XM_011535628.2 | 1 | 804 | XP_011533930.1 | |
| XM_017010585.1 | XP_016866074.1 | |||
| XM_017010586.1 | XP_016866075.1 | |||
| AF032886.1 | AAC39592.1 | |||
| AJ001589.1 | CAA04860.1 | |||
| AK092357.1 | ||||
| AK122861.1 | ||||
| BC020227.1 | AAH20227.1 | |||
| BC021224.2 | AAH21224.1 | |||
| BC068552.1 | AAH68552.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950491 | Chr.6:108574598 - 108653697 on Build GRCh38 | Duplication |
|
| nsv830767 | Chr.6:108476568 - 108667716 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114540116] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map