Assay Details
Target Gene Details
Entrez Gene ID: | 23595 |
Gene Name: | origin recognition complex subunit 3 |
Gene Aliases: |
LAT, LATHEO, ORC3L |
Location: |
Chr.6:87590067-87677824 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6085n100 | Chr.6:87443361 - 87700332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map