Assay Details
Target Gene Details
Entrez Gene ID: | 101929639 |
Gene Name: | B4GALT1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:33166948-33179983 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GALT1-AS1 | NR_108108.1 | |||
| NR_108109.1 | ||||
| NR_108110.1 | ||||
| AB371488.1 | ||||
| HY028011.1 | ||||
| HY043002.1 | ||||
| HY143413.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519369 | Chr.9:33134619 - 33283163 on Build GRCh38 | Gain |
|
| dgv7515n100 | Chr.9:33042019 - 33241605 on Build GRCh38 | Gain |
|
| dgv7516n100 | Chr.9:33121909 - 33278846 on Build GRCh38 | Gain |
|
| esv2762813 | Chr.9:33139254 - 33261075 on Build GRCh38 | Gain |
|

Genomic Map