Assay Details
Target Gene Details
Entrez Gene ID: | 2683 |
Gene Name: | beta-1,4-galactosyltransferase 1 |
Gene Aliases: |
B4GAL-T1, CDG2D, GGTB2, GT1, GTB, beta4Gal-T1 |
Location: |
Chr.9:33110641-33167391 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GALT1 | NM_001497.3 | NP_001488.2 | ||
| XM_005251440.4 | XP_005251497.1 | |||
| A23697.1 | CAA01685.1 | |||
| AK296928.1 | ||||
| AK297541.1 | ||||
| AK312797.1 | ||||
| AK316294.1 | ||||
| BC045773.1 | AAH45773.1 | |||
| BX648438.1 | ||||
| D29805.1 | BAA06188.1 | |||
| HG765099.1 | ||||
| HG765101.1 | ||||
| M13701.1 | AAA35935.1 | |||
| M22921.1 | AAA35936.1 | |||
| U10472.1 | AAA68218.1 | |||
| U10473.1 | AAA68219.1 | |||
| U10474.1 | AAA68220.1 | |||
| X13223.1 | CAA31611.1 | |||
| X14085.1 | CAA32247.1 | |||
| X55415.1 | CAA39073.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv614143 | Chr.9:32583509 - 33141754 on Build GRCh38 | Gain |
|
| dgv7515n100 | Chr.9:33042019 - 33241605 on Build GRCh38 | Gain |
|
| nsv509299 | Chr.9:33048937 - 33173530 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map