Assay Details
Target Gene Details
Entrez Gene ID: | 116225 |
Gene Name: | zinc finger MYND-type containing 19 |
Gene Aliases: |
MIZIP |
Location: |
Chr.9:137582079-137590485 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZMYND19 | NM_138462.2 | NP_612471.1 | ||
| XM_005266052.4 | XP_005266109.1 | |||
| AJ298882.2 | CAC16691.1 | |||
| AX766496.1 | CAE00329.1 | |||
| BC012948.1 | AAH12948.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831762 | Chr.9:137444327 - 137619655 on Build GRCh38 | Loss |
|
| nsv616023 | Chr.9:137478118 - 137708435 on Build GRCh38 | Loss |
|
| nsv831763 | Chr.9:137545379 - 137719569 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map