Assay Details
Target Gene Details
Entrez Gene ID: | 89846 |
Gene Name: | FYVE, RhoGEF and PH domain containing 3 |
Gene Aliases: |
ZFYVE5 |
Location: |
Chr.9:92947319-93036236 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGD3 | NM_001083536.1 | NP_001077005.1 | ||
| NM_001286993.1 | NP_001273922.1 | |||
| NM_033086.2 | NP_149077.2 | |||
| XM_017015274.1 | XP_016870763.1 | |||
| AK000004.1 | BAA92229.1 | |||
| AK225843.1 | ||||
| AK292676.1 | ||||
| AY211386.1 | AAP20645.1 | |||
| BC032429.1 | AAH32429.1 | |||
| BC111054.1 | AAI11055.1 | |||
| BX647829.1 |
Target Gene Details
Entrez Gene ID: | 101927954 |
Gene Name: | uncharacterized LOC101927954 |
Gene Aliases: |
- |
Location: |
Chr.9:93008115-93014944 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927954 | NR_121565.1 | 3 | 1788 | |
| AK126072.1 | 3 | 1787 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526296 | Chr.9:92945910 - 93268110 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map