Assay Details
Target Gene Details
Entrez Gene ID: | 2801 |
Gene Name: | golgin A2 |
Gene Aliases: |
GM130 |
Location: |
Chr.9:128255829-128276007 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLGA2 | NM_004486.4 | NP_004477.3 | ||
| XM_005251930.1 | XP_005251987.1 | |||
| XM_005251931.1 | XP_005251988.1 | |||
| XM_005251932.1 | XP_005251989.1 | |||
| AF248953.1 | AAF65550.1 | |||
| AW583140.1 | ||||
| BC069268.1 | AAH69268.1 | |||
| BX647920.1 | CAI46071.1 |
Target Gene Details
Entrez Gene ID: | 375757 |
Gene Name: | SWI5 homologous recombination repair protein |
Gene Aliases: |
C9orf119, SAE3 |
Location: |
Chr.9:128275379-128288989 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SWI5 | NM_001318092.1 | NP_001305021.1 | ||
| CF129530.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv972808 | Chr.9:128272628 - 128276957 on Build GRCh38 | Duplication |
|
| nsv831728 | Chr.9:128237994 - 128416514 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map