Assay Details
Target Gene Details
Entrez Gene ID: | 100499483 |
Gene Name: | coiled-coil domain containing 180 |
Gene Aliases: |
BDAG1, C9orf174, KIAA1529 |
Location: |
Chr.9:97307628-97377295 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC180 | NM_020893.2 | NP_065944.2 | ||
| AK302965.1 | ||||
| BC047097.1 | AAH47097.2 | |||
| CR749320.1 | CAH18175.1 |
Target Gene Details
Entrez Gene ID: | 57653 |
Gene Name: | LOC100499484-C9orf174 readthrough |
Gene Aliases: |
- |
Location: |
Chr.9:97238426-97377295 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100499484-C9ORF174 | NR_036527.1 | |||
| NR_036528.1 | ||||
| NR_036529.1 |

Genomic Map