Assay Details
Target Gene Details
Entrez Gene ID: | 9933 |
Gene Name: | pumilio RNA binding family member 3 |
Gene Aliases: |
HA-8, HLA-HA8, KIAA0020, PEN, PUF-A, PUF6, XTP5 |
Location: |
Chr.9:2804152-2844130 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PUM3 | NM_014878.4 | NP_055693.4 | ||
| AF490254.1 | AAO85462.1 | |||
| AK225953.1 | ||||
| AK292169.1 | ||||
| AK315531.1 | ||||
| AL832239.1 | ||||
| AL832245.1 | ||||
| BC016137.2 | AAH16137.2 | |||
| CR456957.1 | CAG33238.1 | |||
| D13645.1 | BAA02808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1142547 | Chr.9:2814057 - 2814257 on Build GRCh38 | Tandem Duplication |
|
| esv3619298 | Chr.9:2745732 - 2816795 on Build GRCh38 | Gain |
|
| esv2594391 | Chr.9:2813485 - 2814669 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map