Assay Details
Target Gene Details
Entrez Gene ID: | 2844 |
Gene Name: | G protein-coupled receptor 21 |
Gene Aliases: |
- |
Location: |
Chr.9:123027342-123042751 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR21 | XM_005251933.3 | 2 | 11348 | XP_005251990.1 |
Target Gene Details
Entrez Gene ID: | 23637 |
Gene Name: | RAB GTPase activating protein 1 |
Gene Aliases: |
GAPCENA, TBC1D11 |
Location: |
Chr.9:122931638-123104869 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RABGAP1 | NM_012197.3 | NP_036329.3 | ||
| XM_011518440.2 | XP_011516742.1 | |||
| XM_011518441.2 | XP_011516743.1 | |||
| XM_011518442.2 | XP_011516744.1 | |||
| XM_011518444.2 | XP_011516746.1 | |||
| XM_017014567.1 | XP_016870056.1 | |||
| XM_017014568.1 | XP_016870057.1 | |||
| XM_017014569.1 | XP_016870058.1 | |||
| XM_017014570.1 | XP_016870059.1 | |||
| AB449897.1 | ||||
| AJ011679.2 | CAB40267.2 | |||
| AK131449.1 | BAD18594.1 | |||
| AK294531.1 | ||||
| BC054492.1 | AAH54492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831717 | Chr.9:123018810 - 123172785 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map