Assay Details
Target Gene Details
Entrez Gene ID: | 84814 |
Gene Name: | phospholipid phosphatase 7 (inactive) |
Gene Aliases: |
C9orf67, NET39, PPAPDC3 |
Location: |
Chr.9:131289682-131309262 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLPP7 | NM_032728.3 | NP_116117.3 | ||
| AK027568.1 | BAB55204.1 | |||
| AK075207.1 | BAC11472.1 | |||
| AK315502.1 | ||||
| BC006362.2 | AAH06362.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615536 | Chr.9:131293789 - 131358182 on Build GRCh38 | Loss |
|
| nsv509327 | Chr.9:131241381 - 131448812 on Build GRCh38 | Insertion |
|
| nsv831739 | Chr.9:131145346 - 131310794 on Build GRCh38 | Loss |
|
| nsv466602 | Chr.9:131269410 - 131347804 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map