Assay Details
Target Gene Details
Entrez Gene ID: | 100506422 |
Gene Name: | putative deoxyuridine 5'-triphosphate nucleotidohydrolase-like protein FLJ16323 |
Gene Aliases: |
- |
Location: |
Chr.9:26066675-26126108 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506422 | XM_017014175.1 | XP_016869664.1 |
More Information
Set Membership: |
|

Genomic Map