Assay Details
Target Gene Details
Entrez Gene ID: | 85365 |
Gene Name: | ALG2, alpha-1,3/1,6-mannosyltransferase |
Gene Aliases: |
CDGIi, CMS14, CMSTA3, NET38, hALPG2 |
Location: |
Chr.9:99216425-99221964 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALG2 | NM_033087.3 | NP_149078.1 | ||
| NR_024532.1 | ||||
| AB161356.1 | BAD11905.1 | |||
| AK027417.1 | BAB55099.1 | |||
| AK074704.1 | BAC11150.1 | |||
| AK074988.1 | BAC11337.1 | |||
| AK075172.1 | BAC11449.1 | |||
| AY358697.1 | AAQ89060.1 | |||
| BC017876.1 | AAH17876.1 | |||
| DA076693.1 | ||||
| DB019023.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621214 | Chr.9:98651384 - 99637088 on Build GRCh38 | Gain |
|
| nsv831666 | Chr.9:99112703 - 99256408 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map