Assay Details
Target Gene Details
Entrez Gene ID: | 29126 |
Gene Name: | CD274 molecule |
Gene Aliases: |
B7-H, B7H1, PD-L1, PDCD1L1, PDCD1LG1, PDL1 |
Location: |
Chr.9:5450503-5470567 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD274 | NM_001267706.1 | NP_001254635.1 | ||
| NM_014143.3 | NP_054862.1 | |||
| NR_052005.1 | ||||
| AF177937.1 | AAF25807.1 | |||
| AF233516.1 | AAG18508.1 | |||
| AK001894.1 | BAA91966.1 | |||
| AK300470.1 | ||||
| AK314567.1 | ||||
| AY254342.1 | AAP13470.1 | |||
| AY291313.1 | AAP42144.1 | |||
| AY714881.1 | AAU09634.1 | |||
| BC069381.1 | AAH69381.1 | |||
| BC074984.2 | AAH74984.1 | |||
| BC113734.1 | ||||
| BC113736.1 | ||||
| DQ286582.1 | ABB90152.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv613196 | Chr.9:5345673 - 5514372 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map