Assay Details
Target Gene Details
Entrez Gene ID: | 10999 |
Gene Name: | solute carrier family 27 member 4 |
Gene Aliases: |
ACSVL4, FATP4, IPS |
Location: |
Chr.9:128340517-128361470 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC27A4 | NM_005094.3 | NP_005085.2 | ||
| XM_017014222.1 | XP_016869711.1 | |||
| AF055899.1 | AAD11623.1 | |||
| AK027504.1 | ||||
| AK074831.1 | ||||
| AK290222.1 | ||||
| AK314681.1 | ||||
| BC009959.2 | AAH09959.1 | |||
| BC065003.1 | AAH65003.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831728 | Chr.9:128237994 - 128416514 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76480821] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map