Assay Details
Target Gene Details
Entrez Gene ID: | 347169 |
Gene Name: | olfactory receptor family 1 subfamily B member 1 (gene/pseudogene) |
Gene Aliases: |
OR9-26, OR9-B |
Location: |
Chr.9:122578405-122657626 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR1B1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054800 | Chr.9:122555206 - 122666496 on Build GRCh38 | Loss |
|
| nsv1039785 | Chr.9:122544123 - 122644710 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74360796] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map