Assay Details
Target Gene Details
Entrez Gene ID: | 6619 |
Gene Name: | small nuclear RNA activating complex polypeptide 3 |
Gene Aliases: |
PTFbeta, SNAP50 |
Location: |
Chr.9:15422696-15466751 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNAPC3 | XM_017015056.1 | XP_016870545.1 | ||
| XM_017015057.1 | XP_016870546.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517975 | Chr.9:15286448 - 15534072 on Build GRCh38 | Loss |
|
| esv3619779 | Chr.9:15419572 - 15430882 on Build GRCh38 | Loss |
|
| nsv831520 | Chr.9:15404868 - 15444449 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map