Assay Details
Target Gene Details
Entrez Gene ID: | 114991 |
Gene Name: | zinc finger protein 618 |
Gene Aliases: |
FP13169, NEDD10 |
Location: |
Chr.9:113876282-114056595 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825055 | Chr.9:113875054 - 113878988 on Build GRCh38 | Gain |
|
| esv3621549 | Chr.9:113836116 - 114331078 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map