Assay Details
Target Gene Details
Entrez Gene ID: | 89853 |
Gene Name: | multivesicular body subunit 12B |
Gene Aliases: |
C9orf28, FAM125B |
Location: |
Chr.9:126326844-126507041 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MVB12B | NM_001011703.2 | NP_001011703.1 | ||
| NM_033446.2 | NP_258257.1 | |||
| XM_005252297.1 | XP_005252354.1 | |||
| XM_017015275.1 | XP_016870764.1 | |||
| XM_017015276.1 | XP_016870765.1 | |||
| XM_017015277.1 | XP_016870766.1 | |||
| XM_017015278.1 | XP_016870767.1 | |||
| AK024432.1 | ||||
| AK293740.1 | ||||
| BC028675.1 | AAH28675.1 | |||
| DA339911.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs111779123] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map