Assay Details
Target Gene Details
Entrez Gene ID: | 51450 |
Gene Name: | paired related homeobox 2 |
Gene Aliases: |
PMX2, PRX2 |
Location: |
Chr.9:129665641-129722674 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRRX2 | NM_016307.3 | NP_057391.1 | ||
| AF061970.1 | AAF17708.1 | |||
| BC014645.1 | AAH14645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046477 | Chr.9:129608081 - 129997996 on Build GRCh38 | Gain |
|
| nsv615496 | Chr.9:129654635 - 129711337 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74427757] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map