Assay Details
Target Gene Details
Entrez Gene ID: | 58499 |
Gene Name: | zinc finger protein 462 |
Gene Aliases: |
Zfp462 |
Location: |
Chr.9:106860154-107012258 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF462 | NM_021224.4 | NP_067047.4 | ||
| XM_006717209.3 | XP_006717272.1 | |||
| XM_006717212.3 | XP_006717275.1 | |||
| XM_017014996.1 | 1 | 143 | XP_016870485.1 | |
| XM_017014999.1 | XP_016870488.1 | |||
| AK127803.1 | BAC87142.1 | |||
| BX648965.1 | CAH56168.1 |
More Information
Set Membership: |
|

Genomic Map