Assay Details
Target Gene Details
Entrez Gene ID: | 117531 |
Gene Name: | transmembrane channel like 1 |
Gene Aliases: |
DFNA36, DFNB11, DFNB7 |
Location: |
Chr.9:72521801-72836351 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMC1 | NM_138691.2 | NP_619636.2 | ||
| AF417578.1 | AAL86399.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891682 | Chr.9:72431648 - 72637305 on Build GRCh38 | Loss |
|
| nsv614611 | Chr.9:72406935 - 72532633 on Build GRCh38 | Loss |
|
| nsv831619 | Chr.9:72473018 - 72640449 on Build GRCh38 | Loss |
|
| nsv1048501 | Chr.9:72481579 - 72861027 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map