Assay Details
Target Gene Details
Entrez Gene ID: | 30815 |
Gene Name: | ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6 |
Gene Aliases: |
SIAT7-F, SIAT7F, ST6GALNACVI |
Location: |
Chr.9:127885321-127906725 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST6GALNAC6 | NM_001286999.1 | NP_001273928.1 | ||
| NM_001287000.1 | NP_001273929.1 | |||
| NM_001287002.1 | NP_001273931.1 | |||
| NM_013443.4 | NP_038471.2 | |||
| NR_104629.1 | ||||
| XM_011518610.2 | XP_011516912.1 | |||
| XM_011518611.2 | XP_011516913.1 | |||
| XM_017014658.1 | XP_016870147.1 | |||
| XM_017014659.1 | XP_016870148.1 | |||
| XM_017014660.1 | XP_016870149.1 | |||
| XM_017014661.1 | XP_016870150.1 | |||
| XM_017014662.1 | XP_016870151.1 | |||
| XM_017014663.1 | XP_016870152.1 | |||
| XM_017014665.1 | XP_016870154.1 | |||
| XM_017014666.1 | XP_016870155.1 | |||
| AK023900.1 | BAB14715.1 | |||
| AK226174.1 | ||||
| AK310850.1 | ||||
| AY358672.1 | AAQ89035.1 | |||
| BC006564.2 | AAH06564.1 | |||
| BC007802.2 | AAH07802.1 | |||
| BC016299.2 | AAH16299.1 | |||
| BG699392.1 |
More Information
Set Membership: |
|

Genomic Map