Assay Details
Target Gene Details
Entrez Gene ID: | 8789 |
Gene Name: | fructose-bisphosphatase 2 |
Gene Aliases: |
- |
Location: |
Chr.9:94558714-94593832 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBP2 | NM_003837.3 | NP_003828.2 | ||
| BC113632.1 | ||||
| BC117477.1 | ||||
| CR536483.1 | CAG38722.1 | |||
| Y10812.2 | CAA71772.1 |
Target Gene Details
Entrez Gene ID: | 101928099 |
Gene Name: | prostate cancer associated transcript 7 (non-protein coding) |
Gene Aliases: |
PCAN-R2 |
Location: |
Chr.9:94555045-94568141 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCAT7 | NR_121566.1 | |||
| NR_121567.1 | ||||
| NR_121568.1 | ||||
| NR_121569.1 | ||||
| AK123066.1 | ||||
| BI196098.1 | ||||
| CF146916.1 | ||||
| HY050400.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621113 | Chr.9:94207958 - 94890837 on Build GRCh38 | Gain |
|
| esv2761545 | Chr.9:94263016 - 94594668 on Build GRCh38 | Gain |
|
| dgv1493e214 | Chr.9:94545750 - 94588367 on Build GRCh38 | Gain |
|
| esv3576685 | Chr.9:94439449 - 94658575 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map