Assay Details
Target Gene Details
Entrez Gene ID: | 54801 |
Gene Name: | HAUS augmin like complex subunit 6 |
Gene Aliases: |
Dgt6, FAM29A |
Location: |
Chr.9:19053137-19102942 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAUS6 | NM_001270890.1 | NP_001257819.1 | ||
| NM_017645.4 | NP_060115.3 | |||
| XM_017014830.1 | XP_016870319.1 | |||
| AB046794.1 | BAB13400.1 | |||
| AF533709.1 | AAQ09022.1 | |||
| AF537091.1 | AAN05700.1 | |||
| AK022964.1 | BAB14334.1 | |||
| AK023068.1 | BAB14388.1 | |||
| AK056458.1 | ||||
| AK301852.1 | ||||
| AL832495.1 | ||||
| BC026178.1 | ||||
| BC065935.1 | AAH65935.1 | |||
| BC071625.1 | AAH71625.1 | |||
| BC111041.1 | AAI11042.1 | |||
| BC113981.1 | ||||
| BC114492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161845 | Chr.9:18960488 - 19122709 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs201901061] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map