Assay Details
Target Gene Details
Entrez Gene ID: | 9615 |
Gene Name: | guanine deaminase |
Gene Aliases: |
CYPIN, GUANASE, NEDASIN |
Location: |
Chr.9:72114595-72256045 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDA | NM_001242507.2 | NP_001229436.1 | ||
| XM_011519221.1 | XP_011517523.1 | |||
| XM_017015338.1 | XP_016870827.1 | |||
| AK300418.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573260 | Chr.9:72083033 - 72189116 on Build GRCh38 | Loss |
|
| nsv831618 | Chr.9:71898164 - 72120383 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map