Assay Details
Target Gene Details
Entrez Gene ID: | 27348 |
Gene Name: | torsin family 1 member B |
Gene Aliases: |
DQ1 |
Location: |
Chr.9:129803143-129811424 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOR1B | NM_001317893.1 | NP_001304822.1 | ||
| NM_001317894.1 | NP_001304823.1 | |||
| NM_014506.2 | NP_055321.1 | |||
| NR_133948.1 | ||||
| NR_134027.1 | ||||
| NR_134113.1 | ||||
| XM_005251927.3 | XP_005251984.1 | |||
| AF007872.1 | AAC51733.1 | |||
| AF317129.1 | AAG50271.1 | |||
| AI568476.1 | ||||
| BC015578.2 | AAH15578.1 | |||
| BE315222.1 | ||||
| BP366330.1 | ||||
| CN262366.1 | ||||
| HY036797.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046477 | Chr.9:129608081 - 129997996 on Build GRCh38 | Gain |
|
| esv3621839 | Chr.9:129701704 - 129885823 on Build GRCh38 | Loss |
|
| nsv6729 | Chr.9:129789659 - 129814916 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map