Assay Details
Target Gene Details
Entrez Gene ID: | 1993 |
Gene Name: | ELAV like neuron-specific RNA binding protein 2 |
Gene Aliases: |
HEL-N1, HELN1, HUB |
Location: |
Chr.9:23690099-23850757 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
More Information
Set Membership: |
|

Genomic Map