Assay Details
Target Gene Details
Entrez Gene ID: | 203 |
Gene Name: | adenylate kinase 1 |
Gene Aliases: |
HTL-S-58j |
Location: |
Chr.9:127866480-127886749 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK1 | NM_000476.2 | NP_000467.1 | ||
| NM_001318121.1 | NP_001305050.1 | |||
| NM_001318122.1 | NP_001305051.1 | |||
| XM_017014427.1 | XP_016869916.1 | |||
| XM_017014428.1 | XP_016869917.1 | |||
| AB021871.1 | BAA78534.1 | |||
| AK223501.1 | BAD97221.1 | |||
| AK313758.1 | ||||
| BC001116.2 | AAH01116.1 | |||
| BC100022.1 | AAI00023.1 | |||
| BG772183.1 | ||||
| BT019580.1 | AAV38387.1 | |||
| CR541977.1 | CAG46775.1 | |||
| CR542004.1 | CAG46801.1 | |||
| CR975542.1 | ||||
| DA077967.1 | ||||
| EU794628.1 | ||||
| HY024815.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28616 | Chr.9:127860289 - 127884215 on Build GRCh38 | Gain+Loss |
|
| esv3621795 | Chr.9:127863894 - 127881499 on Build GRCh38 | Gain |
|
| esv3576708 | Chr.9:127866309 - 127882824 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112080972] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map