Assay Details
Target Gene Details
Entrez Gene ID: | 2650 |
Gene Name: | glucosaminyl (N-acetyl) transferase 1, core 2 |
Gene Aliases: |
C2GNT, C2GNT-L, C2GNT1, G6NT, NACGT2, NAGCT2 |
Location: |
Chr.9:76441666-76507416 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCNT1 | NM_001097633.1 | NP_001091102.1 | ||
| NM_001097634.1 | NP_001091103.1 | |||
| NM_001097635.1 | NP_001091104.1 | |||
| NM_001490.4 | NP_001481.2 | |||
| XM_017014620.1 | XP_016870109.1 | |||
| XM_017014621.1 | XP_016870110.1 | |||
| AK313343.1 | ||||
| AL832647.2 | CAD89956.1 | |||
| AY196293.1 | ||||
| AY196294.1 | ||||
| AY196295.1 | ||||
| AY196296.1 | ||||
| BP359964.1 | ||||
| DA479942.1 |
More Information
Set Membership: |
|

Genomic Map