Assay Details
Target Gene Details
Entrez Gene ID: | 9719 |
Gene Name: | ADAMTS like 2 |
Gene Aliases: |
GPHYSD1 |
Location: |
Chr.9:133532164-133575519 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTSL2 | NM_001145320.1 | NP_001138792.1 | ||
| NM_014694.3 | NP_055509.2 | |||
| XM_005272237.3 | XP_005272294.1 | |||
| XM_005272238.3 | XP_005272295.1 | |||
| XM_005272239.2 | XP_005272296.1 | |||
| XM_011519241.2 | XP_011517543.2 | |||
| XM_011519242.2 | XP_011517544.1 | |||
| AB011177.1 | ||||
| BC050544.1 | AAH50544.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| nsv951799 | Chr.9:133473579 - 133617678 on Build GRCh38 | Deletion |
|
| nsv615637 | Chr.9:133532712 - 133539394 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map