Assay Details
Target Gene Details
Entrez Gene ID: | 23349 |
Gene Name: | PHD finger protein 24 |
Gene Aliases: |
KIAA1045 |
Location: |
Chr.9:34810040-34982544 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHF24 | XM_011517827.1 | XP_011516129.1 | ||
| XM_017014553.1 | XP_016870042.1 | |||
| XM_017014554.1 | XP_016870043.1 | |||
| XM_017014555.1 | XP_016870044.1 | |||
| XM_017014556.1 | XP_016870045.1 |
More Information
Set Membership: |
|

Genomic Map